siRNA Details
| Gene Symbol: Antxrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018957.1 | - | 1 | 144 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707274.1 | - | 1 | 137 | Chr. 14 - 34053177 - 34054971 [+] on Build GRCm38 | |

Antxrl
Genomic Map