siRNA Details
| Gene Symbol: 5033428I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017203.1 | - | 1 | 63 | ||
| AK034688.1 | - | 2 | 124 | |||
| AK142377.1 | - | 1 | 63 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB626770.1 | - | 2 | 124 | Chr. 8 - 57332118 - 57333150 [+] on Build GRCm38 | |

5033428I22Rik
Genomic Map