siRNA Details
| Gene Symbol: C330006A16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077073.1 | - | 1 | 1110 | ||
| AK082757.1 | - | 1 | 1113 | |||
| BC020361.1 | - | 1 | 1051 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C330006A16Rik| Gene Symbol: C330006A16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077073.1 | - | 1 | 1110 | ||
| AK082757.1 | - | 1 | 1113 | |||
| BC020361.1 | - | 1 | 1051 | |||