siRNA Details
| Gene Symbol: Gm41819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077070.1 | - | 1 | 87 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB573755.2 | - | 1 | 77 | Chr. 19 - 21992591 - 22020328 [+] on Build GRCm38 | |

Gm41819
Genomic Map