siRNA Details
| Gene Symbol: Gm35585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035376.1 | - | 2 | 1492 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB110862.2 | - | 1 | 623 | Chr. 3 - 127864574 - 127865256 [+] on Build GRCm38 | |

Gm35585
Genomic Map