siRNA Details
| Gene Symbol: BB123696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035379.1 | - | 1 | 187 | ||
| AK035586.1 | - | 1 | 195 | |||
| AK035638.1 | - | 1 | 192 | |||
| AK035711.1 | - | 1 | 188 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB123696| Gene Symbol: BB123696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035379.1 | - | 1 | 187 | ||
| AK035586.1 | - | 1 | 195 | |||
| AK035638.1 | - | 1 | 192 | |||
| AK035711.1 | - | 1 | 188 | |||