siRNA Details
| Gene Symbol: C130026L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038902.1 | - | 135 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C130026L21Rik| Gene Symbol: C130026L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038902.1 | - | 135 | |||