siRNA Details
| Gene Symbol: A330076H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043162.1 | - | 931 | |||
| AK080655.2 | - | 859 | ||||
| AK082028.1 | - | 1959 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330076H08Rik| Gene Symbol: A330076H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043162.1 | - | 931 | |||
| AK080655.2 | - | 859 | ||||
| AK082028.1 | - | 1959 | ||||