siRNA Details
| Gene Symbol: 2810427A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA667402.1 | - | 1 | 321 | ||
| AK013168.1 | - | 1 | 429 | |||
| AV142851.1 | - | 1 | 122 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810427A07Rik| Gene Symbol: 2810427A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA667402.1 | - | 1 | 321 | ||
| AK013168.1 | - | 1 | 429 | |||
| AV142851.1 | - | 1 | 122 | |||