siRNA Details
| Gene Symbol: 4930440I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108098.1 | 5, 6 | 609 | |||
| GenBank mRNA | AK019604.1 | - | 5, 6 | 564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930440I19Rik| Gene Symbol: 4930440I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108098.1 | 5, 6 | 609 | |||
| GenBank mRNA | AK019604.1 | - | 5, 6 | 564 | ||