siRNA Details
| Gene Symbol: 4930440I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108098.1 | 7 | 964 | |||
| GenBank mRNA | AK019604.1 | - | 7 | 919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930440I19Rik| Gene Symbol: 4930440I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108098.1 | 7 | 964 | |||
| GenBank mRNA | AK019604.1 | - | 7 | 919 | ||