siRNA Details
| Gene Symbol: 4930440I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108098.1 | 5 | 557 | |||
| GenBank mRNA | AK019604.1 | - | 5 | 512 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714903.1 | - | 5 | 513 | Chr. 2 - 78051223 - 78188020 [+] on Build GRCm38 | |

4930440I19Rik
Genomic Map