siRNA Details
Gene Symbol: 4930440I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108098.1 | 7 | 888 | |||
GenBank mRNA | AK019604.1 | - | 7 | 843 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930440I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108098.1 | 7 | 888 | |||
GenBank mRNA | AK019604.1 | - | 7 | 843 |