siRNA Details
| Gene Symbol: 4930441H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019606.1 | - | 3 | 449 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714906.1 | - | 3 | 446 | Chr. 7 - 71909697 - 71939191 [+] on Build GRCm38 | |

4930441H08Rik
Genomic Map