siRNA Details
| Gene Symbol: 2410012E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045939.1 | 3 | 294 | |||
| GenBank mRNA | AK010467.1 | - | 3 | 295 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410012E07Rik| Gene Symbol: 2410012E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045939.1 | 3 | 294 | |||
| GenBank mRNA | AK010467.1 | - | 3 | 295 | ||