siRNA Details
Gene Symbol: 4933401H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040540.1 | 5 | 811 | |||
GenBank mRNA | AK016602.1 | - | 4 | 1085 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933401H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040540.1 | 5 | 811 | |||
GenBank mRNA | AK016602.1 | - | 4 | 1085 |