siRNA Details
| Gene Symbol: Gm30918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155516.1 | - | 3 | 657 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI657629.1 | - | 2 | 646 | Chr. 13 - 37542955 - 37548298 [+] on Build GRCm38 | |

Gm30918
Genomic Map