siRNA Details
| Gene Symbol: Gm30918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155516.1 | - | 1, 2 | 62 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY749312.1 | - | 1, 2 | 62 | Chr. 13 - 37541357 - 37548193 [+] on Build GRCm38 | |

Gm30918
Genomic Map