siRNA Details
| Gene Symbol: D730003K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021326.1 | - | 2 | 287 | ||
| BB283416.1 | - | 1 | 270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D730003K21Rik| Gene Symbol: D730003K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021326.1 | - | 2 | 287 | ||
| BB283416.1 | - | 1 | 270 | |||