siRNA Details
| Gene Symbol: D730004N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021329.1 | - | 1 | 88 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY730753.1 | - | 1 | 85 | Chr. 2 - 110710343 - 110710977 [+] on Build GRCm38 | |

D730004N08Rik
Genomic Map