siRNA Details
| Gene Symbol: 2810468A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013383.1 | - | 1 | 384 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712377.1 | - | 1 | 384 | Chr. 11 - 6547177 - 6547685 [+] on Build GRCm38 | |

2810468A05Rik
Genomic Map