siRNA Details
| Gene Symbol: 1700094J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040580.1 | 5 | 880 | |||
| GenBank mRNA | AK007067.1 | - | 5 | 879 | ||
| Gene Symbol: Pcnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG088995.1 | - | 1 | 15 | ||

1700094J05Rik
Genomic Map