siRNA Details
| Gene Symbol: 4931413K12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006915.1 | - | 1 | 469 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706911.1 | - | 1 | 466 | Chr. 11 - 75594260 - 75594737 [-] on Build GRCm38 | |

4931413K12Rik
Genomic Map