siRNA Details
Gene Symbol: Gm41676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053050.1 | - | 1 | 1632 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB526642.2 | - | 1 | 418 | Chr. 18 - 22336038 - 22336817 [-] on Build GRCm38 |
