siRNA Details
| Gene Symbol: Gm41676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053050.1 | - | 1 | 1516 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB526642.2 | - | 1 | 302 | Chr. 18 - 22336038 - 22336817 [-] on Build GRCm38 | |

Gm41676
Genomic Map