siRNA Details
Gene Symbol: E330032C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054493.1 | - | 3 | 348 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB666812.1 | - | 3 | 348 | Chr. 17 - 73252545 - 73258444 [+] on Build GRCm38 |
