siRNA Details
| Gene Symbol: D330050G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040335.1 | 4 | 452 | |||
| NR_040336.1 | 3 | 360 | ||||
| GenBank mRNA | AK083230.1 | - | 4 | 430 | ||
| AK084832.1 | - | 3 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D330050G23Rik| Gene Symbol: D330050G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040335.1 | 4 | 452 | |||
| NR_040336.1 | 3 | 360 | ||||
| GenBank mRNA | AK083230.1 | - | 4 | 430 | ||
| AK084832.1 | - | 3 | 362 | |||