siRNA Details
Gene Symbol: D330050G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040335.1 | 4 | 499 | |||
NR_040336.1 | 3 | 407 | ||||
GenBank mRNA | AK083230.1 | - | 4 | 477 | ||
AK084832.1 | - | 3 | 409 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D330050G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040335.1 | 4 | 499 | |||
NR_040336.1 | 3 | 407 | ||||
GenBank mRNA | AK083230.1 | - | 4 | 477 | ||
AK084832.1 | - | 3 | 409 |