siRNA Details
| Gene Symbol: D330050G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040335.1 | 4 | 1325 | |||
| NR_040336.1 | 3 | 1233 | ||||
| GenBank mRNA | AK083230.1 | - | 4 | 1303 | ||
| AK084832.1 | - | 3 | 1239 | |||
| BB429713.2 | - | 1 | 374 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D330050G23Rik| Gene Symbol: D330050G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040335.1 | 4 | 1325 | |||
| NR_040336.1 | 3 | 1233 | ||||
| GenBank mRNA | AK083230.1 | - | 4 | 1303 | ||
| AK084832.1 | - | 3 | 1239 | |||
| BB429713.2 | - | 1 | 374 | |||