siRNA Details
| Gene Symbol: D330050G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040335.1 | 2 | 192 | |||
| GenBank mRNA | AK083230.1 | - | 2 | 170 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB653363.1 | - | 2 | 167 | Chr. 2 - 116900177 - 116911679 [+] on Build GRCm38 | |

D330050G23Rik
Genomic Map