siRNA Details
| Gene Symbol: Slc28a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085680.1 | - | 3 | 818 | ||
| BC037061.1 | - | 2 | 392 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc28a3| Gene Symbol: Slc28a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085680.1 | - | 3 | 818 | ||
| BC037061.1 | - | 2 | 392 | |||