siRNA Details
| Gene Symbol: 6430584L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046179.1 | 4 | 1351 | |||
| GenBank mRNA | AK032533.1 | - | 4 | 1352 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6430584L05Rik| Gene Symbol: 6430584L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046179.1 | 4 | 1351 | |||
| GenBank mRNA | AK032533.1 | - | 4 | 1352 | ||