siRNA Details
| Gene Symbol: Dhrs13os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053376.1 | - | 2 | 1546 | ||
| AK084880.1 | - | 1 | 1716 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhrs13os| Gene Symbol: Dhrs13os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053376.1 | - | 2 | 1546 | ||
| AK084880.1 | - | 1 | 1716 | |||