siRNA Details
| Gene Symbol: Gpatch8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084888.1 | - | 1 | 635 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB481873.2 | - | 1 | 411 | Chr. 11 - 102555284 - 102555893 [-] on Build GRCm38 | |

Gpatch8
Genomic Map