siRNA Details
| Gene Symbol: 5730507A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020490.1 | - | 1 | 294 | ||
| AK051924.1 | - | 1 | 2706 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5730507A11Rik| Gene Symbol: 5730507A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020490.1 | - | 1 | 294 | ||
| AK051924.1 | - | 1 | 2706 | |||