siRNA Details
| Gene Symbol: C330013F16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045455.1 | 6 | 587 | |||
| GenBank mRNA | AK133643.1 | - | 6 | 588 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C330013F16Rik| Gene Symbol: C330013F16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045455.1 | 6 | 587 | |||
| GenBank mRNA | AK133643.1 | - | 6 | 588 | ||