siRNA Details
| Gene Symbol: Gm38413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044041.1 | - | 9 | 1700 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB273420.2 | - | 1 | 63 | Chr. 5 - 99730854 - 99735303 [-] on Build GRCm38 | |

Gm38413
Genomic Map