siRNA Details
| Gene Symbol: Mob3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041672.1 | - | 2534 | |||
| AK164429.1 | - | 2923 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mob3a| Gene Symbol: Mob3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041672.1 | - | 2534 | |||
| AK164429.1 | - | 2923 | ||||