siRNA Details
| Gene Symbol: 4931408D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040298.1 | 4 | 1392 | |||
| NR_040299.1 | 3 | 1211 | ||||
| GenBank mRNA | AK016444.1 | - | 4 | 1187 | ||
| AK040817.1 | - | 3 | 1133 | |||
| AK168860.1 | - | 3 | 1204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931408D14Rik| Gene Symbol: 4931408D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040298.1 | 4 | 1392 | |||
| NR_040299.1 | 3 | 1211 | ||||
| GenBank mRNA | AK016444.1 | - | 4 | 1187 | ||
| AK040817.1 | - | 3 | 1133 | |||
| AK168860.1 | - | 3 | 1204 | |||