siRNA Details
| Gene Symbol: 1700008C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005755.1 | - | 4, 5 | 569 | ||
| BY705915.1 | - | 4, 5 | 567 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700008C04Rik| Gene Symbol: 1700008C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005755.1 | - | 4, 5 | 569 | ||
| BY705915.1 | - | 4, 5 | 567 | |||