siRNA Details
Gene Symbol: Dcdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138215.1 | - | 1 | 78 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY723817.1 | - | 1 | 78 | Chr. 2 - 106226454 - 106336145 [+] on Build GRCm38 |
