siRNA Details
| Gene Symbol: Plekha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041498.1 | - | 480 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB637826.1 | - | 2 | 475 | Chr. 8 - 25037057 - 25038414 [+] on Build GRCm38 | |

Plekha2
Genomic Map