siRNA Details
Gene Symbol: 1700006J14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005678.1 | - | 2 | 185 | ||
AK011105.1 | - | 2 | 177 | |||
BC098228.1 | - | 2 | 183 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700006J14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005678.1 | - | 2 | 185 | ||
AK011105.1 | - | 2 | 177 | |||
BC098228.1 | - | 2 | 183 |