siRNA Details
| Gene Symbol: 1700006J14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005678.1 | - | 1 | 53 | ||
| AK011105.1 | - | 1 | 45 | |||
| BC098228.1 | - | 1 | 51 | |||
| Gene Symbol: Hmga2-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037996.1 | 1 | 42 | |||
| GenBank mRNA | AK033703.1 | - | 1 | 43 | ||

1700006J14Rik
Genomic Map