siRNA Details
| Gene Symbol: 8430416G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038171.1 | - | 1, 2 | 241 | ||
| Gene Symbol: Gm4013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033452.1 | 1, 2 | 251 | |||

8430416G17Rik
Genomic Map