siRNA Details
| Gene Symbol: BC049265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158865.1 | - | 990 | |||
| BC030475.1 | - | 890 | ||||
| BC049265.1 | - | 879 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC049265| Gene Symbol: BC049265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158865.1 | - | 990 | |||
| BC030475.1 | - | 890 | ||||
| BC049265.1 | - | 879 | ||||