siRNA Details
| Gene Symbol: Hnrnpab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019387.1 | - | 395 | |||
| AK047205.1 | - | 2210 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnrnpab| Gene Symbol: Hnrnpab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019387.1 | - | 395 | |||
| AK047205.1 | - | 2210 | ||||