siRNA Details
| Gene Symbol: Gm12802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084081.1 | - | 2 | 469 | ||
| BB657329.1 | - | 2 | 469 | |||
| BG807815.1 | - | 2 | 254 | |||
| CX568958.1 | - | 2 | 426 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12802| Gene Symbol: Gm12802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084081.1 | - | 2 | 469 | ||
| BB657329.1 | - | 2 | 469 | |||
| BG807815.1 | - | 2 | 254 | |||
| CX568958.1 | - | 2 | 426 | |||