siRNA Details
| Gene Symbol: Gm19626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044311.1 | - | 2523 | |||
| AK149278.1 | - | 1505 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19626| Gene Symbol: Gm19626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044311.1 | - | 2523 | |||
| AK149278.1 | - | 1505 | ||||