siRNA Details
| Gene Symbol: 4930429D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040699.1 | 7 | 662 | |||
| GenBank mRNA | AK015233.1 | - | 7 | 663 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930429D17Rik| Gene Symbol: 4930429D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040699.1 | 7 | 662 | |||
| GenBank mRNA | AK015233.1 | - | 7 | 663 | ||